Gene Transcripts

Gene Symbol
Sec61b
Entrez Gene ID
66212
Gene Name
Sec61 beta subunit
Gene Aliases
1190006C12Rik, AI326121, AW122942
Chromosome Location
Chr. 4: 47474661 - 47483233 on Build GRCm38
UniGene ID
Mm.20188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024171.2 NP_077133.1 4 379
GenBank mRNA AK004505.1 - 4 380
AK008739.1 - 4 383
BC002089.1 - 4 351 3487410
BC081445.1 - 4 347 6412192

View Full Product Details