Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025485.3 | NP_079761.1 | 2 | 341 | |
GenBank mRNA | AK003836.1 | - | 1 | 109 | |
AK013925.1 | - | 2 | 322 | ||
AK154600.1 | - | 2 | 322 | ||
BC037653.1 | - | 2 | 318 | ||
BC051198.1 | - | 2 | 321 | 1396237 | |
CJ183743.1 | - | 2 | 341 |