Gene Transcripts
- Gene Symbol
- NUDC
- Entrez Gene ID
- 10726
- Gene Name
- nuclear distribution C, dynein complex regulator
- Gene Aliases
- HNUDC, MNUDC, NPD011
- Chromosome Location
- Chr. 1: 26911128 - 26946871 on Build GRCh38
- UniGene ID
- Hs.263812
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006600.3 |
NP_006591.1
|
2, 3 |
280 |
|
XM_011540529.1 |
XP_011538831.1
|
2, 3 |
180 |
|
XM_011540530.1 |
XP_011538832.1
|
2, 3 |
418 |
|
GenBank mRNA |
AB019408.1 |
- |
2, 3 |
227 |
|
AF086922.1 |
- |
2, 3 |
243 |
|
AF100760.1 |
- |
2, 3 |
243 |
|
AF125465.1 |
- |
2, 3 |
251 |
|
AF130736.1 |
- |
2, 3 |
194 |
|
AF241788.1 |
- |
2, 3 |
228 |
|
AL136725.1 |
- |
2, 3 |
255 |
|
BC002399.2 |
- |
2, 3 |
192 |
2822167 |
BC003132.1 |
- |
2, 3 |
200 |
2958251 |
BC006147.1 |
- |
2, 3 |
226 |
3954220 |
BC007280.1 |
- |
2, 3 |
269 |
3343588 |
BC015153.1 |
- |
2, 3 |
268 |
3883328 |
BC021139.1 |
- |
2, 3 |
241 |
4871007 |
BP348144.1 |
- |
2, 3 |
280 |
|
View Full Product Details