Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029979.3 | NP_084255.2 | 1 | 314 | |
GenBank mRNA | AB060155.1 | - | 1 | 258 | |
AF145374.1 | - | 1 | 298 | ||
AK003000.1 | - | 1 | 273 | ||
AK020775.1 | - | 1 | 275 | ||
AK031803.1 | - | 1 | 268 | ||
AK032635.1 | - | 1 | 287 | ||
AK075877.1 | - | 1 | 285 | ||
AK129288.1 | - | 1 | 259 | ||
AK163749.1 | - | 1 | 250 | ||
BC049105.1 | - | 1 | 264 | 6418046 | |
BC053494.1 | - | 1 | 179 | ||
BC145783.1 | - | 1 | 308 | 40131086 | |
BC145809.1 | - | 1 | 308 | 40131112 |