Gene Transcripts
- Gene Symbol
- Trim35
- Entrez Gene ID
- 66854
- Gene Name
- tripartite motif-containing 35
- Gene Aliases
- 0710005M05Rik, A430106H13Rik, AW046487, HLS5, Mair, NC8, mKIAA1098
- Chromosome Location
- Chr. 14: 66297025 - 66311424 on Build GRCm38
- UniGene ID
- Mm.43871
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029979.3 |
NP_084255.2
|
2, 3 |
620 |
|
GenBank mRNA |
AB060155.1 |
- |
2, 3 |
564 |
|
AF145374.1 |
- |
2, 3 |
604 |
|
AK003000.1 |
- |
2, 3 |
579 |
|
AK032635.1 |
- |
2, 3 |
593 |
|
AK075877.1 |
- |
2, 3 |
591 |
|
AK146541.1 |
- |
1, 2 |
71 |
|
AK163749.1 |
- |
2, 3 |
556 |
|
BC049105.1 |
- |
2, 3 |
570 |
6418046 |
BC053494.1 |
- |
2, 3 |
485 |
|
BC145783.1 |
- |
2, 3 |
614 |
40131086 |
BC145809.1 |
- |
2, 3 |
614 |
40131112 |
View Full Product Details