Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170745.1 | NP_001164216.1 | 7 | 1241 | |
NM_001170746.1 | NP_001164217.1 | 8 | 1406 | ||
NM_015823.3 | NP_056638.1 | 7 | 1241 | ||
GenBank mRNA | AK039336.1 | - | 4 | 226 | |
AK122349.1 | - | 6 | 701 | ||
AK147530.1 | - | 7 | 1242 | ||
BC059005.1 | - | 7 | 898 | 6847238 |