Gene Transcripts

Gene Symbol
Map3k6
Entrez Gene ID
53608
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
Ask2, MAPKKK6, MEKK6
Chromosome Location
Chr. 4: 133239669 - 133252929 on Build GRCm38
UniGene ID
Mm.36640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016693.5 NP_057902.5 16 2377
GenBank mRNA AB021861.4 - 16 2375
AK204377.1 - 2 131
BC120565.1 - 16 2304 40129488
BC125577.1 - 16 2304 40130092

View Full Product Details