Gene Transcripts

Gene Symbol
Mtx2
Entrez Gene ID
53375
Gene Name
metaxin 2
Gene Aliases
1500012G02Rik
Chromosome Location
Chr. 2: 74825812 - 74876748 on Build GRCm38
UniGene ID
Mm.292613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016804.4 NP_058084.3 9, 10 818
GenBank mRNA AB041562.1 - 9, 10 810
AF053550.1 - 9, 10 791
AK005233.1 - 9, 10 750
AK083055.1 - 780
AK087933.1 - 9, 10 806
AK168000.1 - 9, 10 811
AK168021.1 - 9, 10 781
AK168045.1 - 9, 10 810
AK169048.1 - 9, 10 814
AK207939.1 - 2, 3 128
AK212612.1 - 2, 3 128
AK216047.1 - 2, 3 128
BC006641.1 - 9, 10 768 3483318

View Full Product Details