Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001169114.1 | NP_001162585.1 | 2 | 609 | |
NM_016888.5 | NP_058584.3 | 2 | 732 | ||
GenBank mRNA | AF092050.2 | - | 2 | 510 | |
AK160088.1 | - | 2 | 701 | ||
AY043479.1 | - | 2 | 690 | ||
BC009075.1 | - | 2 | 609 | 2654354 |