Gene Transcripts

Gene Symbol
Rabep1
Entrez Gene ID
54189
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
Rab5ep, rabaptin-5, rabaptin-5alpha
Chromosome Location
Chr. 11: 70844723 - 70943105 on Build GRCm38
UniGene ID
Mm.7087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291141.1 NP_001278070.1 5 747
NM_001291142.1 NP_001278071.1 5 756
NM_001291143.1 NP_001278072.1 7 947
NM_019400.3 NP_062273.2 6 876
GenBank mRNA AB001750.1 - 7 783
AF248489.1 - 5 542
AF248490.1 - 5 533
AK019413.1 - 6 850
AK134587.1 - 6 876
AK137236.1 - 6 841
BC009814.1 - 6 872
BC080274.1 - 6 849
BC129853.1 - 6 695 40109688
BC129854.1 - 6 695 40109689
D86066.1 - 6 712

View Full Product Details