Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019519.2 | NP_062392.1 | 15 | 1874 | |
GenBank mRNA | AF127656.1 | - | 16 | 1722 | |
AF127657.1 | - | 15 | 1665 | ||
AF127658.1 | - | 15 | 1874 | ||
AF127659.1 | - | 15 | 1874 | ||
AF127660.1 | - | 15 | 1819 | ||
AF127661.1 | - | 14 | 1762 | ||
AF127662.1 | - | 16 | 1793 | ||
AK002625.1 | - | 16 | 1656 | ||
AK146917.1 | - | 16 | 1745 | ||
BC012214.1 | - | 14 | 1908 | 3967218 |