Gene Transcripts

Gene Symbol
Rbx1
Entrez Gene ID
56438
Gene Name
ring-box 1
Gene Aliases
1500002P15Rik, AA517855, ROC1
Chromosome Location
Chr. 15: 81466316 - 81476369 on Build GRCm38
UniGene ID
Mm.29405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019712.3 NP_062686.1 5 680
GenBank mRNA AK004114.1 - 5 644
BC027396.1 - 5 629 4952242
BC056992.1 - 5 623 6818827

View Full Product Details