Gene Transcripts

Gene Symbol
Snd1
Entrez Gene ID
56463
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
AL033314, Tudor-SN
Chromosome Location
Chr. 6: 28480315 - 28935162 on Build GRCm38
UniGene ID
Mm.439987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019776.2 NP_062750.2 2 380
GenBank mRNA AB021491.1 - 2 337
AK034914.1 - 2 328
AK088028.1 - 2 339
AK133837.1 - 2 386
AK159837.1 - 2 345
AK161591.1 - 2 334
AK162432.1 - 2 321
AK167576.1 - 2 370
AK204079.1 - 1 46
BB613775.1 - 2 380
BC007126.1 - 2 327 3592548
BM944959.1 - 1 58

View Full Product Details