Gene Transcripts

Gene Symbol
Gmeb1
Entrez Gene ID
56809
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
1110050A04Rik, AI256615, AI481278
Chromosome Location
Chr. 4: 132220376 - 132261549 on Build GRCm38
UniGene ID
Mm.103489
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122992.1 NP_001116464.1 3 235
NM_020273.2 NP_064669.2 2 172
GenBank mRNA AF210433.1 - 1 133
AK134349.1 - 2 173
BC046983.1 - 3 206 6477130
BY732360.1 - 3 210
DV660282.1 - 1 90

View Full Product Details