Gene Transcripts
- Gene Symbol
- Vps25
- Entrez Gene ID
- 28084
- Gene Name
- vacuolar protein sorting 25 (yeast)
- Gene Aliases
- 1110020N13Rik, AW107467, D11Wsu68e
- Chromosome Location
- Chr. 11: 101253611 - 101259547 on Build GRCm38
- UniGene ID
- Mm.301020
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284412.1 |
NP_001271341.1
|
2, 3 |
248 |
|
NM_001284414.1 |
NP_001271343.1
|
2, 3 |
248 |
|
NM_026776.4 |
NP_081052.2
|
2, 3 |
248 |
|
GenBank mRNA |
AK002500.1 |
- |
2, 3 |
210 |
|
AK003715.2 |
- |
2, 3 |
220 |
|
AK003877.1 |
- |
2, 3 |
206 |
|
AK082933.1 |
- |
2, 3 |
227 |
|
AK145811.1 |
- |
2, 3 |
219 |
|
AK150822.1 |
- |
2, 3 |
230 |
|
AK162230.1 |
- |
2, 3 |
221 |
|
AK166008.1 |
- |
2, 3 |
227 |
|
BC029181.1 |
- |
2, 3 |
212 |
4240500 |
View Full Product Details