Gene Transcripts

Gene Symbol
Uba1y
Entrez Gene ID
22202
Gene Name
ubiquitin-activating enzyme, Chr Y
Gene Aliases
A1s9Y-1, Sby, Ube-2, Ube1ay, Ube1y, Ube1y-1, Ube1y1
Chromosome Location
Chr. Y: 818612 - 844224 on Build GRCm38
UniGene ID
Mm.422949
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011667.2 NP_035797.1 5 489
GenBank mRNA AF150963.1 - 5 489
AK030031.1 - 5 488

View Full Product Details