Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011678.2 | NP_035808.2 | 2 | 297 | |
GenBank mRNA | AK138838.1 | - | 2 | 232 | |
AK143582.1 | - | 2 | 232 | ||
AK149964.1 | - | 2 | 255 | ||
AK169372.1 | - | 2 | 276 | ||
AK169933.1 | - | 2 | 267 | ||
AK171271.1 | - | 2 | 267 | ||
AK171633.1 | - | 2 | 250 | ||
AK214431.1 | - | 1 | 93 | ||
AK220512.1 | - | 2 | 254 | ||
BC066180.1 | - | 2 | 207 | ||
BC066865.1 | - | 2 | 207 | ||
BY322979.1 | - | 2 | 297 | ||
L00681.1 | - | 2 | 260 |