Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011842.3 | NP_035972.3 | 5 | 763 | |
GenBank mRNA | AF159259.1 | - | 5 | 638 | |
AK077548.1 | - | 5 | 745 | ||
AK088158.1 | - | 5 | 676 | ||
AK147099.1 | - | 5 | 742 | ||
AK149834.1 | - | 5 | 809 | ||
AK157717.1 | - | 5 | 667 | ||
AK159979.1 | - | 5 | 739 | ||
AK180159.1 | - | 3 | 190 | ||
AK183116.1 | - | 3 | 190 | ||
AK206505.1 | - | 2 | 160 | ||
BC079847.1 | - | 5 | 763 | 6419411 |