Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011942.1 | NP_036072.1 | 9 | 709 | |
GenBank mRNA | AB009653.1 | - | 9 | 709 | |
AK003689.1 | - | 9 | 676 | ||
AK075590.1 | - | 9 | 701 | ||
AK089112.1 | - | 9 | 701 | ||
AK167478.1 | - | 9 | 696 | ||
BC068120.1 | - | 9 | 735 | 5714650 |