Gene Transcripts

Gene Symbol
Mapk14
Entrez Gene ID
26416
Gene Name
mitogen-activated protein kinase 14
Gene Aliases
CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha
Chromosome Location
Chr. 17: 28691342 - 28748405 on Build GRCm38
UniGene ID
Mm.311337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168508.1 NP_001161980.1 11, 12 1422
NM_001168513.1 NP_001161985.1 11, 12 1023
NM_001168514.1 NP_001161986.1 12, 13 1467
NM_011951.3 NP_036081.1 11, 12 1422
GenBank mRNA AK089059.1 - 11, 12 1336
AK133684.1 - 11, 12 1335
AK151348.1 - 11, 12 1024
AK153025.1 - 11, 12 1024
BC012235.1 - 11, 12 1422 4195842
BC018227.1 - 12, 13 1364
BC085303.1 - 11, 12 1398 5389457
D83073.1 - 11, 12 1253
U10871.1 - 11, 12 1320

View Full Product Details