Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013760.4 | NP_038788.2 | 3 | 582 | |
GenBank mRNA | AK005475.1 | - | 3 | 583 | |
BC036125.1 | - | 2 | 446 | ||
BC042713.1 | - | 2 | 476 | 4039558 | |
BC096676.1 | - | 3 | 581 | 963849 | |
CT010304.1 | - | 2 | 443 |