Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159502.1 | NP_001152974.1 | 2 | 424 | |
NM_020494.3 | NP_065240.2 | 2 | 258 | ||
GenBank mRNA | AF214732.1 | - | 2 | 238 | |
AK011012.1 | - | 2 | 202 | ||
AK018862.1 | - | 2 | 240 | ||
AK036538.1 | - | 2 | 222 | ||
AK134339.1 | - | 2 | 236 | ||
AK149810.1 | - | 2 | 209 | ||
AK150778.1 | - | 2 | 210 | ||
AK152551.1 | - | 2 | 210 | ||
AK152928.1 | - | 2 | 210 | ||
AK166360.1 | - | 2 | 216 | ||
AK172557.1 | - | 2 | 209 | ||
BC043721.1 | - | 2 | 226 | ||
BC055048.1 | - | 2 | 188 | 6484820 | |
BC055317.1 | - | 2 | 424 | 5696084 | |
CJ141277.1 | - | 2 | 258 |