Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022813.3 | NP_073724.1 | 9 | 1825 | |
GenBank mRNA | AK161002.1 | - | 9 | 1824 | |
AK171766.1 | - | 9 | 1814 | ||
BC014751.1 | - | 9 | 1803 | 4235556 | |
CA979044.1 | - | 1 | 634 |