Gene Transcripts

Gene Symbol
Def6
Entrez Gene ID
23853
Gene Name
differentially expressed in FDCP 6
Gene Aliases
2410003F05Rik, 6430538D02Rik, AV094905, Ibp, Slat, Slat2, Slat6
Chromosome Location
Chr. 17: 28207778 - 28228608 on Build GRCm38
UniGene ID
Mm.204731
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027185.3 NP_081461.2 3 337
GenBank mRNA AF329497.1 - 3 272
AK038050.1 - 3 308
AK088460.1 - 3 286
AY241695.1 - 3 272
BC120500.1 - 3 337 40129423
BC131953.1 - 3 337 40130230
X96705.1 - 3 229

View Full Product Details