Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021588.2 | NP_067599.1 | 3 | 832 | |
GenBank mRNA | BC070511.1 | - | 3 | 832 | |
FQ214690.1 | - | 3 | 864 | ||
FQ214898.1 | - | 3 | 863 | ||
FQ214986.1 | - | 3 | 864 | ||
FQ215026.1 | - | 3 | 864 | ||
FQ215032.1 | - | 3 | 863 | ||
FQ215043.1 | - | 3 | 863 | ||
FQ215217.1 | - | 3 | 862 | ||
FQ215441.1 | - | 3 | 863 | ||
FQ215511.1 | - | 3 | 862 | ||
FQ215553.1 | - | 3 | 863 | ||
FQ215560.1 | - | 3 | 862 | ||
FQ215564.1 | - | 3 | 862 | ||
FQ215642.1 | - | 3 | 873 | ||
FQ215802.1 | - | 3 | 864 | ||
FQ215880.1 | - | 3 | 863 | ||
FQ215982.1 | - | 3 | 863 | ||
FQ216127.1 | - | 3 | 862 | ||
FQ216280.1 | - | 3 | 870 | ||
FQ216306.1 | - | 3 | 865 | ||
FQ216335.1 | - | 3 | 864 | ||
FQ216380.1 | - | 3 | 863 | ||
FQ216504.1 | - | 3 | 863 | ||
FQ216709.1 | - | 3 | 862 | ||
FQ216731.1 | - | 3 | 862 | ||
FQ217450.1 | - | 3 | 863 | ||
FQ217496.1 | - | 3 | 862 | ||
FQ218027.1 | - | 3 | 863 | ||
FQ223624.1 | - | 3 | 862 | ||
FQ223672.1 | - | 3 | 859 | ||
FQ223774.1 | - | 3 | 864 | ||
FQ224523.1 | - | 3 | 863 | ||
FQ224833.1 | - | 1 | 354 | ||
FQ224856.1 | - | 3 | 863 | ||
FQ224909.1 | - | 3 | 864 | ||
FQ224921.1 | - | 3 | 860 | ||
FQ224975.1 | - | 3 | 863 |