Gene Transcripts
- Gene Symbol
- Grk1
- Entrez Gene ID
- 81760
- Gene Name
- G protein-coupled receptor kinase 1
- Gene Aliases
- Rhok
- Chromosome Location
- Chr. 16: 81153438 - 81169128 on Build Rnor_6.0
- UniGene ID
- Rn.10548
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031096.1 |
NP_112358.1
|
4 |
1221 |
|
GenBank mRNA |
U63971.1 |
- |
4 |
1221 |
|
View Full Product Details