Gene Transcripts

Gene Symbol
Tmsb4x
Entrez Gene ID
81814
Gene Name
thymosin beta 4, X-linked
Gene Aliases
Chromosome Location
Chr. X: 28593405 - 28617267 on Build Rnor_6.0
UniGene ID
Rn.201192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031136.1 NP_112398.1 3 242
GenBank mRNA BC058137.1 - 3 256
BC084720.1 - 3 231
BC097308.1 - 3 241
FQ211846.1 - 3 249
FQ211950.1 - 3 252
FQ213262.1 - 3 249
FQ214330.1 - 3 249
FQ217154.1 - 3 249
FQ217532.1 - 3 249
FQ217617.1 - 3 249
FQ217622.1 - 3 249
FQ220985.1 - 3 249
FQ221085.1 - 3 249
FQ221124.1 - 3 249
FQ221299.1 - 3 249
FQ221505.1 - 3 249
FQ221785.1 - 3 249
FQ221964.1 - 3 249
FQ222006.1 - 3 249
FQ222011.1 - 3 249
FQ222095.1 - 3 249
FQ222205.1 - 3 249
FQ222251.1 - 3 249
FQ222334.1 - 3 249
FQ222360.1 - 3 243
FQ222463.1 - 3 249
FQ222519.1 - 3 249
FQ222755.1 - 3 249
FQ223266.1 - 3 246
FQ223325.1 - 3 249
FQ228380.1 - 3 249
FQ228401.1 - 3 249
FQ228412.1 - 3 249
FQ228477.1 - 3 249
FQ229728.1 - 3 250
K01334.1 - 2 210
M26759.1 - 2 216
M34043.1 - 3 242

View Full Product Details