Gene Transcripts

Gene Symbol
Fgf4
Entrez Gene ID
116499
Gene Name
fibroblast growth factor 4
Gene Aliases
Hst, Hst1
Chromosome Location
Chr. 1: 218025701 - 218027235 on Build Rnor_6.0
UniGene ID
Rn.81223
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053809.1 NP_446261.1 3 574
XM_006230640.2 XP_006230702.1 3 704
GenBank mRNA AB079673.1 - 3 574

View Full Product Details