Gene Transcripts

Gene Symbol
Rasgrp4
Entrez Gene ID
170668
Gene Name
RAS guanyl releasing protein 4
Gene Aliases
Chromosome Location
Chr. 1: 88078350 - 88094331 on Build Rnor_6.0
UniGene ID
Rn.213071
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130824.2 NP_570837.2 4 438
GenBank mRNA AF465263.1 - 4 438
AF465264.1 - 4 438

View Full Product Details