Gene Transcripts

Gene Symbol
Grin3b
Entrez Gene ID
170796
Gene Name
glutamate receptor, ionotropic, N-methyl-D-aspartate 3B
Gene Aliases
GluN3B, NMDAR3B, NR3B
Chromosome Location
Chr. 7: 12764993 - 12771310 on Build Rnor_6.0
UniGene ID
Rn.162906
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133308.2 NP_579842.2 2 934
GenBank mRNA AF440691.2 - 2 934

View Full Product Details