Gene Transcripts

Gene Symbol
Cryba2
Entrez Gene ID
286925
Gene Name
crystallin, beta A2
Gene Aliases
Chromosome Location
Chr. 9: 82151056 - 82154266 on Build Rnor_6.0
UniGene ID
Rn.204241
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173140.1 NP_775163.1 5 660
GenBank mRNA AY158891.1 - 5 660

View Full Product Details