Gene Transcripts

Gene Symbol
Rabep1
Entrez Gene ID
54190
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
Chromosome Location
Chr. 10: 57462284 - 57581520 on Build Rnor_6.0
UniGene ID
Rn.5876
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019124.1 NP_061997.1 5, 6 702
XM_006246795.2 XP_006246857.1 5, 6 865
XM_006246796.2 XP_006246858.1 5, 6 865
XM_008767861.1 XP_008766083.1 5, 6 1496
GenBank mRNA D85844.1 - 5, 6 702
U70777.1 - 5, 6 849

View Full Product Details