Gene Transcripts

Gene Symbol
TREH
Entrez Gene ID
11181
Gene Name
trehalase
Gene Aliases
TRE, TREA
Chromosome Location
Chr. 11: 118658232 - 118679672 on Build GRCh38
UniGene ID
Hs.129712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301065.1 NP_001287994.1 3 277
NM_007180.2 NP_009111.2 3 277
GenBank mRNA AB000824.1 - 3 287
AK091981.1 - 2 154
AK223140.1 - 3 251
AK223143.1 - 3 255
AK309565.1 - 3 251
BC109206.2 - 3 271 40006501
DA626089.1 - 2 194

View Full Product Details