Gene Transcripts

Gene Symbol
Cyp2c23
Entrez Gene ID
83790
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 23
Gene Aliases
Chromosome Location
Chr. 1: 263778510 - 263803150 on Build Rnor_6.0
UniGene ID
Rn.2184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031839.3 NP_114027.2 2 241
GenBank mRNA BC078707.1 - 2 241
U04733.1 - 2 222

View Full Product Details