Gene Transcripts

Gene Symbol
Cyp2c23
Entrez Gene ID
83790
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 23
Gene Aliases
Chromosome Location
Chr. 1: 263778510 - 263803150 on Build Rnor_6.0
UniGene ID
Rn.2184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031839.3 NP_114027.2 6, 7 998
GenBank mRNA BC078707.1 - 6, 7 998
FQ209508.1 - 4, 5 597
FQ218733.1 - 1, 2 125
FQ218882.1 - 3, 4 347
U04733.1 - 6, 7 979

View Full Product Details