Gene Transcripts

Gene Symbol
Mterf
Entrez Gene ID
85261
Gene Name
mitochondrial transcription termination factor
Gene Aliases
Mterf1
Chromosome Location
Chr. 4: 27365310 - 27372374 on Build Rnor_6.0
UniGene ID
Rn.64629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053499.1 NP_445951.1 3 406
XM_006236069.2 XP_006236131.1 4 521
XM_006236070.2 XP_006236132.1 4 518
XM_006236071.2 XP_006236133.1 3 393
XM_006236072.2 XP_006236134.1 3 390
XM_006236073.2 XP_006236135.1 2 344
GenBank mRNA AJ292524.1 - 3 406

View Full Product Details