Gene Transcripts
- Gene Symbol
- Ppp2r2c
- Entrez Gene ID
- 117256
- Gene Name
- protein phosphatase 2, regulatory subunit B, gamma
- Gene Aliases
- Pr52
- Chromosome Location
- Chr. 14: 78679214 - 78763175 on Build Rnor_6.0
- UniGene ID
- Rn.54550
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057116.1 |
NP_476457.1
|
|
1331 |
|
GenBank mRNA |
D38261.1 |
- |
|
1331 |
|
View Full Product Details