Gene Transcripts

Gene Symbol
Khsrp
Entrez Gene ID
171137
Gene Name
KH-type splicing regulatory protein
Gene Aliases
Marta1
Chromosome Location
Chr. 9: 10013792 - 10023670 on Build Rnor_6.0
UniGene ID
Rn.92643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133602.1 NP_598286.1 3 362
XM_006244292.2 XP_006244354.1 3 500
GenBank mRNA AF308818.1 - 3 362

View Full Product Details