Gene Transcripts

Gene Symbol
Akr7a2
Entrez Gene ID
171445
Gene Name
aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase)
Gene Aliases
Aiar
Chromosome Location
Chr. 5: 157759448 - 157768473 on Build Rnor_6.0
UniGene ID
Rn.8548
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134407.1 NP_599234.1 7 1103
GenBank mRNA AB037424.1 - 7 1103
AF503514.1 - 7 1141
AJ271883.1 - 7 1106
BC061816.1 - 7 1106 5598724
FQ230711.1 - 4 512
FQ231883.1 - 4 512

View Full Product Details