Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001713.1 | NP_001001713.1 | 2 | 127 | |
NM_001317740.1 | NP_001304669.1 | 2 | 380 | ||
NM_001317741.1 | NP_001304670.1 | 2 | 270 | ||
NM_001317742.1 | NP_001304671.1 | 2 | 96 | ||
NM_007341.2 | NP_031367.1 | 2 | 356 | ||
GenBank mRNA | AW962501.1 | - | 2 | 87 | |
BC006371.2 | - | 2 | 307 | 4138452 | |
BG742101.1 | - | 2 | 270 | ||
BI819819.1 | - | 2 | 95 | ||
BM474020.1 | - | 2 | 143 | ||
BM697281.1 | - | 2 | 356 | ||
BM701533.1 | - | 2 | 380 | ||
BT006994.1 | - | 2 | 302 | ||
X93498.1 | - | 2 | 319 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317744.1 | NP_001304673.1 | 4 | 545 | |
GenBank mRNA | CD514470.1 | - | 4 | 360 | |
FY213114.1 | - | 4 | 361 | ||
KM576720.1 | - | 4 | 387 |