Gene Transcripts

Gene Symbol
Prkaa1
Entrez Gene ID
65248
Gene Name
protein kinase, AMP-activated, alpha 1 catalytic subunit
Gene Aliases
AMPKalpha1
Chromosome Location
Chr. 2: 54857688 - 54893404 on Build Rnor_6.0
UniGene ID
Rn.87789
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019142.2 NP_062015.2 2 248
GenBank mRNA FN804412.1 - 2 226
U40819.1 - 2 215

View Full Product Details