Gene Transcripts
- Gene Symbol
- Hpse
- Entrez Gene ID
- 64537
- Gene Name
- heparanase
- Gene Aliases
- Hep, Hpa
- Chromosome Location
- Chr. 14: 10534358 - 10576686 on Build Rnor_6.0
- UniGene ID
- Rn.6392
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022605.1 |
NP_072127.1
|
12 |
2936 |
|
GenBank mRNA |
AF184967.1 |
- |
12 |
2936 |
|
View Full Product Details