Gene Transcripts

Gene Symbol
Bag6
Entrez Gene ID
94342
Gene Name
BCL2-associated athanogene 6
Gene Aliases
Bat3
Chromosome Location
Chr. 20: 5125324 - 5138084 on Build Rnor_6.0
UniGene ID
Rn.203343
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033968.1 NP_001029140.1 6 618
NM_053609.2 NP_446061.2 6 776
XM_006256082.1 XP_006256144.1 6 680
XM_006256083.1 XP_006256145.1 6 680
XM_006256084.1 XP_006256146.1 5 626
XM_006256085.1 XP_006256147.1 6 680
XM_006256086.2 XP_006256148.1 6 816
XM_006256087.2 XP_006256149.1 6 893
XM_006256088.1 XP_006256150.1 6 680
XM_006256089.1 XP_006256151.1 6 680
XM_006256090.1 XP_006256152.1 5 626
XM_006256091.1 XP_006256153.1 5 626
XM_006256092.1 XP_006256154.1 6 680
XM_006256093.2 XP_006256155.1 5 764
XM_006256094.1 XP_006256156.1 6 680
GenBank mRNA AB018791.1 - 6 732
BC100141.1 - 6 618 7315623
FQ223705.1 - 5 739

View Full Product Details