Gene Transcripts

Gene Symbol
Wfdc1
Entrez Gene ID
171112
Gene Name
WAP four-disulfide core domain 1
Gene Aliases
ps20
Chromosome Location
Chr. 19: 52313771 - 52332721 on Build Rnor_6.0
UniGene ID
Rn.3193
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133581.1 NP_598265.1 7 763
GenBank mRNA AF037272.1 - 7 763
BC063152.1 - 7 766 6887007
FQ213083.1 - 7 748
FQ216345.1 - 7 757
FQ216415.1 - 7 755
FQ219846.1 - 7 758
FQ219958.1 - 7 996
FQ220334.1 - 7 994
FQ229404.1 - 7 748

View Full Product Details