Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308373.1 | NP_001295302.1 | 6 | 957 | |
NM_012181.4 | NP_036313.3 | 6 | 960 | ||
XM_011527863.1 | XP_011526165.1 | 6 | 1018 | ||
XM_011527864.1 | XP_011526166.1 | 5 | 849 | ||
GenBank mRNA | AK026952.1 | - | 1548 | ||
AK300571.1 | - | 6 | 931 | ||
AK313363.1 | - | 6 | 891 | ||
AY225339.1 | - | 5 | 821 | ||
AY278607.1 | - | 6 | 884 | ||
BC009966.1 | - | 5 | 664 | 4299915 | |
BX538124.1 | - | 6 | 938 | ||
BX647405.1 | - | 6 | 937 | ||
BX647720.1 | - | 6 | 895 | ||
GQ372970.1 | - | 3 | 457 |