Gene Transcripts
- Gene Symbol
- Dync1i1
- Entrez Gene ID
- 29564
- Gene Name
- dynein cytoplasmic 1 intermediate chain 1
- Gene Aliases
- Dnci1, Dncic1
- Chromosome Location
- Chr. 4: 30807704 - 31121095 on Build Rnor_6.0
- UniGene ID
- Rn.11273
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019234.1 |
NP_062107.1
|
4 |
394 |
|
GenBank mRNA |
X66845.1 |
- |
4 |
394 |
|
View Full Product Details