Gene Transcripts

Gene Symbol
Gjb5
Entrez Gene ID
29586
Gene Name
gap junction protein, beta 5
Gene Aliases
Cx31.1
Chromosome Location
Chr. 5: 145422034 - 145425035 on Build Rnor_6.0
UniGene ID
Rn.16074
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019241.1 NP_062114.1 1 680
XM_008763990.1 XP_008762212.1 2 924

View Full Product Details