Gene Transcripts

Gene Symbol
Gja6
Entrez Gene ID
54256
Gene Name
gap junction protein, alpha 6
Gene Aliases
cx33, cxn-33
Chromosome Location
Chr. X: 35618584 - 35622951 on Build Rnor_6.0
UniGene ID
Rn.209042
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019308.1 NP_062181.1 1 410
XM_006256894.2 XP_006256956.1 2 787

View Full Product Details