Gene Transcripts

Gene Symbol
Prl3c1
Entrez Gene ID
58937
Gene Name
Prolactin family 3, subfamily c, member 1
Gene Aliases
Prlpi, Prlpj
Chromosome Location
Chr. 17: 42812517 - 42818305 on Build Rnor_6.0
UniGene ID
Rn.3248
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031316.1 NP_112606.1 4 485
GenBank mRNA AB019119.1 - 4 485
AB019945.1 - 4 476
AF150741.1 - 4 485
AF234638.1 - 4 476

View Full Product Details