Gene Transcripts

Gene Symbol
Scp2
Entrez Gene ID
25541
Gene Name
sterol carrier protein 2
Gene Aliases
NSLIPTR, SCPx
Chromosome Location
Chr. 5: 127647934 - 127735703 on Build Rnor_6.0
UniGene ID
Rn.31887
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138508.4 NP_612517.2 2 104
GenBank mRNA BC081713.1 - 2 117
M57453.1 - 2 104
M62763.1 - 2 131
X60654.1 - 2 84

View Full Product Details